Learn More
Bochem™ Stainless Steel/PTFE Coated Standard Spoon Spatulas
Click to view available options
Length (Metric):
210 mm
230 mm
missing translation for 'unitSize'
Each
Specifications
Specifications
| Quantity | 1 Ea. |
| Product Type | Spoon Spatula |
| Diameter (Metric) Outer | 4mm |
| Material | Stainless Steel/PTFE Coated |
| Length (Metric) | 230 mm |