Learn More
Grant Instruments™ Stainless Steel Perforated Base Tray
Click to view available options
For Use With (Equipment):
DMU12 & SAP12
DMU19, SAP18/26
DMU5, SAP2/5, JBN5
LSB12
OLS26/LSB18
SAP34
Unit Size:
Each
Description
Required if flat-bottomed flasks are to be placed directly on the base of the bath. Grant Instruments™ Stainless Steel Replacement Base Trays promote thermal convection in the bath.
- For use with SBB Aqua Plus and LSB Series Aqua Pro Series Unstirred Water Bath
- Choose from a vartiety of sizes.
Specifications
Specifications
| For Use With (Equipment) | OLS26/LSB18 |
| Type | SBT 26 Stainless Steel Perforated Base Tray |
| Material | Stainless Steel |