Description
- Stainless steel
- Available for a variety of test sieves, based upon diameter
Specifications
Specifications
| Quantity | 1 Ea. |
| Type | Sieve Cover |
| Certifications/Compliance | ISO 3310 and ASTM E 11 |
| Cover Material | Stainless Steel |
| Diameter (Metric) Outer | 100 mm |