Learn More
RETSCH Stainless-Steel Lid for Test Sieves
Shop All Retsch Products
missing translation for 'orderingAttributeHoverText'
Diameter (Metric) Outer:
100 mm
150 mm
200 mm
305 mm
315 mm
missing translation for 'unitSize'
Each
Description
- Stainless steel
- Available for a variety of test sieves, based upon diameter
Specifications
Specifications
| Quantity | 1 Ea. |
| Type | Sieve Lid |
| Certifications/Compliance | ISO 3310 and ASTM E 11 |
| Cover Material | Stainless Steel |
| Diameter (Metric) Outer | 150 mm |