Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for IN750plus, 15 mm
Shop All Memmert Products
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Description
- Accessory
- May affect temperature distribution
- Constructed of stainless steel
- Rim height: 15 mm
Specifications
Specifications
| For Use With (Equipment) | IN750plus Incubator |
| Material | Stainless Steel |
| Type | Drip Tray |
| Height (Metric) | Rim: 15 mm |