Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for SF110plus, 15 mm
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
SF110plus Sterilizer
Unit Size:
Each
Description
- Accessory
- May affect temperature distribution
- Constructed of stainless steel
- Rim height: 15 mm
Specifications
Specifications
| For Use With (Equipment) | SF110plus Sterilizer |
| Product Type | Drip Tray |
| Height (Metric) Tray | Rim: 15 mm |
| Material | Stainless Steel |