Learn More
Memmert™ Stainless Steel Slide-In Drip Tray
Product Code. 17190045 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
HPP1060eco
HPP110eco
HPP260eco
HPP410eco
HPP750eco
IPP1060eco
IPP110eco
IPP260eco
IPP410eco
IPP750eco
UN110pa
Unit Size:
Each
For Use With (Application):
Do not use with Modified Chamber
Reinforced Chamber Option K1
Product Code. 17190045
Brand: Memmert™ IPP1060ecoB32599
Only null left
Add to basket
Add to basket
Stainless steel slide-in bottom drip tray
  • May affect the temperature distribution
  • 15 mm rim
Specifications
Description Sliding drip tray, 15 mm rim
For Use With (Equipment) IPP1060eco
Type Drip Tray
Length (Metric) 15 mm rim