Learn More
Bochem™ Stainless Steel Stand Base
Click to view available options
Length (Metric):
210 mm
250 mm
300 mm
315 mm
Width (Metric):
130 mm
150 mm
160 mm
200 mm
Unit Size:
Each
Description
- Expanded, flat and heavy feet for even more stability
- 1 adjustable foot
- Anti-slide rubber protection
- Thread M10
Specifications
Specifications
| Material | Stainless Steel (18/10) |
| Height (Metric) | 3 mm |
| Length (Metric) | 315 mm |
| Width (Metric) | 200 mm |
| Product Type | Stand Base |
| Quantity | 1 Pk. |