Learn More
Bochem™ Stainless Steel Stand Base
Click to view available options
Length (Metric):
210 mm
250 mm
300 mm
315 mm
Width (Metric):
130 mm
150 mm
160 mm
200 mm
Unit Size:
Each
Description
- Expanded, flat and heavy feet for stability
- 1 adjustable foot
- Anti-slide rubber protection
- Thread M10
Specifications
Specifications
| Material | Stainless Steel (18/10) |
| Height (Metric) | 3 mm |
| Length (Metric) | 210 mm |
| Width (Metric) | 130 mm |
| Includes | M10 thread |
| Product Type | Stand Base |
| Shape | Flat |
| Thread Size | M10 |
| Quantity | 1 Ea. |
| Weight (Metric) | 1.6 kg |