Learn More
Amsta Rostfritt Stainless Steel Transport Container - Acid Proof With Skirtband, Without Lid
Product Code. p-200113425
Click to view available options
Capacity (Metric):
20 L
25 L
30 L
50 L
75 L
100 L
150 L
Unit Size:
Each
Product Code. 30285051
Brand: Amsta Rostfritt 516081000
Highest quality stainless steel transport container - acid proof. Made of 1 mm stainless steel (EN1.4404). Intended for antibiotics, vaccines, chemicals and liquids. Manufactured in Sweden.
  • Highest quality in stainless steel.
  • Acid proof 316L/1.4404.
  • Straight cylindrical vessel in highly polished finish with stylish design.
Specifications
Product Type Pail
Capacity (Metric) 100 L
Color Silver
Material Stainless Steel (316L)