Learn More
Amsta Rostfritt Stainless Steel Transport Container Without Skirtband, Without Lid
Product Code. 30284887
Click to view available options
Capacity (Metric):
20 L
25 L
30 L
50 L
75 L
100 L
Unit Size:
Each
Product Code. 30284887
Brand: Amsta Rostfritt 50609750
Highest quality stainless steel transport container. Manufactured in Sweden.
  • Stainless steel 304 /1.4301
  • Straight cylindrical vessel in highly polished finish with stylish design.
Specifications
Product Type Pail
Capacity (Metric) 75 L
Color Silver
Material Stainless Steel