Learn More
Wuertt Celluloid™ Stainless Steel Tube Rack
Click to view available options
Positions:
48
Unit Size:
Each
Specifications
Specifications
| Cell Dividers | 4 x 12 |
| Product Type | Tube Rack |
| Array | 4 x 12 |
| Holds | Tubes (18 mm) |
| Material | Stainless Steel |
| Positions | 48 |
| For Use With (Equipment) | For Test Tube |
| Quantity | 1 Pack |
| Length (Metric) | 250 mm |
| Width (Metric) | 90 mm |
| Show More |