Learn More
RUBIS Long Tail Tweezers
Product Code. p-8008444
Encompass_Preferred
Click to view available options
Length (Metric):
255 mm
Length (English):
10.04 in.
Unit Size:
Each
Product Code. 10143442
Brand: RUBIS 232/10 SA
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Long Tail Tweezers - Antimagnetic/Anti-acid SS, Serrated Handle
  • Long Tail Tweezers
  • 100% Made in Switzerland
  • Anti-acid/Antimagnetic Stainless Steel
  • Serrated handles
  • Serrated tips for better grip
  • Blunt tips
  • Suitable for cryogenics applications
  • Strong
Specifications
Tip Style Straight, Round, Blunt, Serrated
Length (English) 10.04 in.
Material Anti-acid, Anti-magnetic Stainless Steel
Length (Metric) 255 mm
Product Type Long Tail Tweezers
Quantity (No. per Case) Each