Learn More
Trajan Scientific and Medical™ Stainless Steel Union Tees
Click to view available options
Description:
1/16 x 1/16 x 1/16 in.
1/8 x 1/16 x 1/16 in.
Union Tee
Unit Size:
Each
Description
- Low dead volume
- High temperature applicationsLeak free connections
Specifications
Specifications
| Type | Union Tee |
| Diameter (English) Outer | 1/4 x 1/16 x 1/16 in. |
| Description | Union Tee |
| Material | Stainless Steel |