Learn More
Trajan Scientific and Medical™ Stainless Steel Union Tees
Product Code. 15299113
Click to view available options
Description:
1/16 x 1/16 x 1/16 in.
1/8 x 1/16 x 1/16 in.
Union Tee
Unit Size:
Each
Product Code. 15299113
Brand: Trajan Scientific and Medical™ 1034180
Only null left
Add to basket
Add to basket
Give low dead-volume connections for optimum chromatography.
  • Low dead volume
  • High temperature applicationsLeak free connections
Specifications
Type Union Tee
Description 1/8 x 1/16 x 1/16 in.
Material Stainless Steel