Learn More
Gestigkeit Harry™ Stainless Steel Universal Bath
Brand:  Gestigkeit Harry™ PZ26-4
Additional Details : Weight : 11.00000kg
Product Code. 10507132
  • € 1830.00 / Each
Estimated Shipment: 24-06-2024
to see stock.
Specifications
Stainless Steel
20°C to 400°C
Fluid-, oil-, air- or sandbath for continuous operation
280 mm
230 V
5 L
Digital
310 x 310 x 280 mm
6.8 kg