Learn More
WTW™ Stainless Steel USP Kit 2
Product Code. 10339743
Click to view available options
Accuracy (TDS):
±2%
Unit Size:
Each
Product Code. 10339743
Brand: WTW™ 300568
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For conductivity measuring systems, there are different check methods:The system including measuring cell can be calibrated with suitable traceably certifed standard solutions. An adjustment is usually not necessary due to the long-term stability of the used electrode materials. Special kits contain all necessary parts to test conductivity measuring systems according to pharamcopeia guidelines.
TRUSTED_SUSTAINABILITY
Specifications
Accuracy (TDS) ±2%
Type UPS KIT 2
For Use With (Application) Conductivity Measuring According to Pharmacopoeia
Material Stainless Steel