Learn More
Ohaus™ Stainless Steel Weight, OIML F1, OH
Shop All Ohaus Products
Click to view available options
Weight (Metric):
100 g
200 g
1 kg
Unit Size:
Each
Description
- Stainless steel construction
- Include a weight case
- Single body design without adjusting cavity
Specifications
Specifications
| Description | Weight, 1kg, OIML F1, OH |
| Classification | OIML F1 Weights |
| Material | Stainless Steel |
| Weight (Metric) | 1 kg |
| Tolerance | Meets or exceeds OIML F1 |