Learn More
Wuertt Celluloid™ Stainless Steel Wire Basket
Product Code. 10372661
Change view
Click to view available options
Depth (Metric):
100 mm
200 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Depth (Metric) unitSize
10372661 100 mm Each
10519051 200 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10372661 Supplier Wuertt Celluloid™ Supplier No. 5200113
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Specifications
Material Stainless Steel
For Use With (Application) Storage and Transport
Quantity 1 Each
Depth (Metric) 100 mm
Product Type Basket
Height (Metric) 100 mm
Width (Metric) 100 mm