Learn More
Wuertt Celluloid™ Stainless Steel Wire Basket
Product Code. 10519051
Change view
Click to view available options
Depth (Metric):
100 mm
200 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Depth (Metric) Unit Size
10519051 200 mm Each
10372661 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 10519051 Supplier Wuertt Celluloid™ Supplier No. 5201313
  • ONLINE EXCLUSIVE PRICE
    € 116.63
    € 70.69 / Pack of 1
    Save € 45.94
    39% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Specifications
Material Stainless Steel
For Use With (Application) Storage and Transport
Quantity 1 Each
Depth (Metric) 200 mm
Product Type Basket
Height (Metric) 200 mm
Width (Metric) 200 mm