Learn More
Linker Industrietechnik™ Stainless Steel Wire Test Tube Wire Frame
Brand:  Linker Industrietechnik™ 516.182.070-0406
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10035050
  • € 84.20 / Each
Estimated Shipment: 17-05-2023
to see stock.
Specifications
Tubes (16 mm)
24
18 x 18 x 70mm
16 mm
Wire, Stainless
Test Tube Rack
16 mm