missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAM-1 Antibody (29C678) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB100-56533JF635
This item is not returnable.
View return policy
Description
STAM-1 Monoclonal antibody specifically detects STAM-1 in Human samples. It is validated for Western Blot
Specifications
| STAM-1 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| DKFZp686J2352, HSE1 homolog, signal transducing adapter molecule 1, signal transducing adaptor molecule (SH3 domain and ITAM motif) 1, STAM1STAM-1 | |
| This antibody was developed against a synthetic peptide corresponding to amino acids 149-165 of human STAM. | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication, Signal Transduction | |
| 8027 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot | |
| 29C678 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction