Learn More
STAM Mouse, anti-Human, Clone: 1F5C2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68628-1-IG-150UL
This item is not returnable.
View return policy
Description
The STAM antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human STAM. This antibody recognizes human, mouse, rat antigen. The STAM antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| STAM | |
| 1F5C2 | |
| DKFZp686J2352, STAM, STAM 1, STAM1 | |
| 150 μL | |
| 8027 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG2b |