Learn More
STAM Mouse, anti-Human, Clone: 1F5C2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68628-1-IG-150UL
 View more versions of this product
Product Code. 18099435
  • € 410.00 / 150µL
Description
The STAM antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human STAM. This antibody recognizes human, mouse, rat antigen. The STAM antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
STAM
1F5C2
DKFZp686J2352, STAM, STAM 1, STAM1
150 μL
8027
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat
150 kDa
IgG2b