missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAM2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-2838
This item is not returnable.
View return policy
Description
STAM2 Polyclonal specifically detects STAM2 in Human samples. It is validated for Western Blot, Flow Cytometry, Peptide ELISA.
Specifications
| STAM2 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| DKFZp564C047, HBP, Hrs-binding protein, HSE1 homolog, signal transducing adapter molecule 2, signal transducing adaptor molecule (SH3 domain and ITAM motif) 2, STAM-2, STAM2A, STAM2B, STAM-like protein containing SH3 and ITAM domains 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10254 | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/mL, Flow Cytometry 10 ug/mL, Peptide ELISA Detection limit 1:8000 | |
| NP_005834.3 | |
| STAM2 | |
| Peptide with sequence C-QSIDPTDSKPDSQD corresponding to internal region according to NP_005834.3. | |
| 0.1 mg | |
| Primary | |
| No cross-reaction expected with STAM1. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction