missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STAM2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518500
This item is not returnable.
View return policy
Description
This antibody is predicted to react with canine and mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
The protein encoded by this gene is closely related to STAM, an adaptor protein involved in the downstream signaling of cytokine receptors, both of which contain a SH3 domain and the immunoreceptor tyrosine-based activation motif (ITAM). Similar to STAM, this protein acts downstream of JAK kinases, and is phosphorylated in response to cytokine stimulation. This protein and STAM thus are thought to exhibit compensatory effects on the signaling pathway downstream of JAK kinases upon cytokine stimulation.
Specifications
| STAM2 | |
| Polyclonal | |
| Unconjugated | |
| STAM2 | |
| 1200004O12Rik; 5730456G07Rik; BB145015; DKFZp564C047; Hbp; HGNC:11358; Hrs binding protein; hrs-binding protein; HSE1 homolog; signal transducing adapter molecule 2; signal transducing adaptor molecule (SH3 domain and ITAM motif) 2; signal transducing adaptor molecule 2; STAM2; STAM-2; STAM2A; STAM2B; STAM-like protein containing SH3 and ITAM domains 2 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 10254 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O75886 | |
| STAM2 | |
| Synthetic peptide sequence (QSIDPTDSKPDSQD) corresponding to the internal amino acids of STAM2 (aa 305-319). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction