missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STAMBP Monoclonal Antibody (K56003_18H5), Invitrogen™
Mouse Monoclonal Antibody
€ 480.00 - € 846.00
Specifications
| Antigen | STAMBP |
|---|---|
| Clone | K56003_18H5 |
| Concentration | 1 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30194214
|
Invitrogen™
MA556128 |
200 μL |
€ 846.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30202242
|
Invitrogen™
MA555748 |
50 μL |
€ 480.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: KSAVIPEKKD TVKKLKEIAF PKAEELKAEL LKRYTKEYTE YNEEKKKEAE ELARNMAIQQ ELEKEKQRVA QQKQQQLEQE QFHAFEEMIR NQELEKERLK IVQEFGKVDP GLGGPLVPDL EKPSLDVFPT LTVSSIQPSD CHTTVRPAKP PVVDRSLKPG ALSNSESIPT
STAMBP Monoclonal antibody specifically detects STAMBP in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
| STAMBP | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 5330424L14Rik; 5730422L11Rik; AMSH; associated molecule with the SH3 domain of STAM; AW107289; Endosome-associated ubiquitin isopeptidase; MGC126516; MGC126518; MICCAP; mKIAA4198; Stam binding protein; STAM-binding protein; Stambp; testicular secretory protein Li 54 | |
| Recombinant human STAMBP (immunogen range is 81-250). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Stambp |
| K56003_18H5 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O95630 | |
| 10617 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title