missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STAMBP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115965
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total STAMBP.
Cytokine-mediated signal transduction in the JAK-STAT cascade requires the involvement of adaptor molecules. One such signal-transducing adaptor molecule contains an SH3 domain that is required for induction of MYC and cell growth. The protein encoded by this gene binds to the SH3 domain of the signal-transducing adaptor molecule, and plays a critical role in cytokine-mediated signaling for MYC induction and cell cycle progression. Multiple alternatively spliced transcript variants encoding the same protein isoform have been found for this gene.
Specifications
| STAMBP | |
| Polyclonal | |
| Unconjugated | |
| Stambp | |
| 5330424L14Rik; 5730422L11Rik; AMSH; associated molecule with the SH3 domain of STAM; AW107289; Endosome-associated ubiquitin isopeptidase; MGC126516; MGC126518; MICCAP; mKIAA4198; Stam binding protein; STAM-binding protein; Stambp; testicular secretory protein Li 54 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10617, 171565, 70527 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O95630, Q8R424, Q9CQ26 | |
| Stambp | |
| A synthesized peptide derived from human STAMBP(Accession O95630), corresponding to amino acid residues S368-V418. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction