missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAMP2/STEAP4 Antibody (418714), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4626I
This item is not returnable.
View return policy
Description
STAMP2/STEAP4 Monoclonal antibody specifically detects STAMP2/STEAP4 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| STAMP2/STEAP4 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 418714 | |
| Flow Cytometry, CyTOF-ready | |
| EC 1.16.1, EC 1.16.1.-, FLJ23153, Six-transmembrane epithelial antigen of prostate 4, SixTransMembrane protein of prostate 2, STAMP2DKFZp666D049, STEAP family member 4, TIARPsix transmembrane prostate protein 2, TNFAIP9, Tumor necrosis factor, alpha-induced protein 9metalloreductase STEAP4, tumor necrosis-alpha-induced adipose-related protein | |
| NS0 mouse myeloma cell line transfected with human STAMP2, Met1-His459, Accession # EAW76907 | |
| 0.1 mL | |
| Cancer | |
| 79689 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction