Learn More
Standard Buffer Solution pH 4, SI Analytics™
Product Code. p-8014348 Shop All SI Analytics Products
Click to view available options
Quantity:
1000 mL
250 mL
60 Ampules
Unit Size:
1 L
250mL
Each
Product Code. 10222451
Brand: SI Analytics 285138213
SI Analytics™ Standard Buffer Solution pH 4 resists changes in pH when small quantities of an acid are added to it.
  • Includes manufacturer certificate
  • Mixture, consisting of the following components: Water, potassium hydrogenphthalate
  • Store tigthly sealed at temperatures between 15°C and 25°C
  • The product does not contain any relevant quantities of materials with critical values that have to be monitored at the workplace
  • Rinse off of bigger amounts into drains or the aquatic environment may lead to decreased pH-values. A low pH-value harms aquatic organisms. In the dilution of the use-level the pH-value is considerably increased, so that after the use of the product the aqueous waste, emptied into drains, is only low water-dangerous
  • Absorb with liquid-binding material (sand, diatomite, acid binders, universal binders, sawdust)
TRUSTED_SUSTAINABILITY
Specifications
Boiling Point 100°C
Color Yellow
Density 1.01g/cm3
pH 4
Physical Form Liquid
Quantity 60 Ampules
Formulation Water, potassium hydrogenphthalate
Odor Odorless
Packaging FIOLAX™ Ampules
Sterility Sterile
Vapor Pressure 23hPa
Solubility Information Fully miscible in water