Learn More
Standard Buffer Solution, SI Analytics™
Product Code. 11994709 Shop All SI Analytics Products
Click to view available options
Quantity:
250 mL
60 Ampules
Unit Size:
250mL
Each
Product Code. 11994709
Brand: SI Analytics 285138024
SI Analytics™ Standard Buffer Solution pH 9.18 resists changes in pH when small quantities of an alkali are added to it.
  • Includes manufacturer certificate
  • Mixture, consisting of the following components: Water, disodium tetraborate decahydrate
  • The product does not contain any relevant quantities of materials with critical values that have to be monitored at the workplace
  • Absorb with liquid-binding material (sand, diatomite, acid binders, universal binders, sawdust)
TRUSTED_SUSTAINABILITY
Specifications
Boiling Point 100°C
Color Colorless
Density 1g/cm3
pH 9.18
Physical Form Liquid
Quantity 250 mL
Formulation Water, disodium tetraborate decahydrate
Odor Odorless
Packaging DURAN™ Bottle
Sterility Sterile
Vapor Pressure 23hPa
Solubility Information Fully miscible in water