Learn More
Tenak™ Standard Rack for Microtiterplates, for Chest Freezers, 25 mm Shelf Height
Shop All TENAK Products
Click to view available options
Holds:
Microtiterplates
Microtiterplates (21)
Microtiterplates (23)
Microtiterplates (25)
Microtiterplates (27)
Unit Size:
Each
Description
- For use in chest freezers
- Racks come with a locking rod to secure the cryoboxes
- Constructed of stainless steel
Specifications
Specifications
| Includes | Locking rod |
| Holds | Microtiterplates (21) |
| Material | Stainless Steel |
| For Use With (Equipment) | Chest Frseezers |
| Dimensions (L x W x H) | 135 x 92 x 561 mm |
| Height (Metric) | 25 mm |