Learn More
Tenak™ Standard Rack for Microtiterplates, for Upright Freezers, 16 mm Shelf Height
Shop All TENAK ProductsDescription
- For use in upright freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of stainless steel
Specifications
Specifications
| Material | Stainless Steel |
| For Use With (Equipment) | Upright Freezers |
| Holds | Microtiterplates (80) |
| Tube Array | 16 x 5 |
| Dimensions (L x W x H) | 450 x 135 x 272 mm |
| Height (Metric) | 16 mm |