Learn More
Edmund Bühler GmbH™ TiMix 5 Standard Rack System
Product Code. 10601532
Click to view available options
For Use With (Equipment):
microplates
Unit Size:
Each
Product Code. 10601532
Brand: Edmund Bühler GmbH™ 0052 101
Only null left
Add to basket
Add to basket
For 8 standard microplates, microwell or deepwell plates, or other plates of the same size.
  • Fits 8 standard microplates, microwell or deepwell plates, or other plates of the same size
  • For use with TiMix 5 Microplate Shakers
Order Info
dimType
Specifications
Product Type Rack System
For Use With (Equipment) microplates