Learn More
Tenak™ Standard Rack for Upright Freezer, 50 mm Box Height
Standard Rack fits into any upright freezer and can be used with all standard cryoboxes.
€ 95.40 - € 128.00
Specifications
Material | Stainless Steel |
---|---|
For Use With (Equipment) | Upright Freezers |
Height (Metric) | 50 mm (Box) |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
11561815
|
Tenak™
TE23120 |
Boxes (9) |
€ 95.40
Each |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||||
15230664
|
Tenak™
TE23136 |
Boxes (24) |
€ 119.00
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
15230674
|
Tenak™
TE23156 |
Boxes (30) |
€ 128.00
Each |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||||
15270664
|
Tenak™
TE23150 |
Boxes (15) |
€ 109.00
Each |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||||
15280654
|
Tenak™
TE23130 |
Boxes (12) |
€ 97.70
Each |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||||
Description
- Measure the inside height and depth of your freezer when choosing the right rack
- Racks can be used with all freezer brands
- Constructed of stainless steel
Specifications
Stainless Steel | |
Upright Freezers | |
50 mm (Box) |