Learn More
Tenak™ Standard Rack for Upright Freezer, 75 mm Box Height
Product Code. 15210684 Shop All TENAK Products
Click to view available options
Holds:
Boxes
Array:
2 x 3
2 x 4
2 x 5
3 x 2
3 x 3
3 x 5
4 x 3
4 x 4
4 x 5
5 x 4
Unit Size:
Each
Product Code. 15210684
Brand: Tenak™ TE23174
Only null left
Add to basket
Add to basket
Standard Rack fits into any upright freezer and can be used with all standard cryoboxes.
  • Measure the inside height and depth of your freezer when choosing the right rack
  • Racks can be used with all freezer brands
  • Constructed of stainless steel
Specifications
Array 4 x 3
Holds Boxes
Material Stainless Steel
Positions 12
For Use With (Equipment) Upright Freezers
Dimensions (L x W x H) 422 x 139 x 320 mm
Height (Metric) 75 mm (Box)