Learn More
Bochem™ Standard Retort Clamps
Corked finger. Bochem™ Standard Retort Clamps have a round jaw.
Brand:  Bochem™ 5461
Additional Details : Weight : 0.24300kg
 View more versions of this product
Product Code. 10333431
  • € 14.60 / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Round jaw
  • Cast iron chromium plated or steel zincked
  • DIN 12894
Specifications
Specifications
1 Ea.
Cast Iron, Chromium-Plated
150 mm
Clamp
25 mm
Round