Learn More
Bochem™ Standard Retort Clamps
Corked finger. Bochem™ Standard Retort Clamps have a round jaw.
Brand:  Bochem™ 5464
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 10769601
  • € 23.10 / Each
Estimated Shipment: 14-05-2024
to see stock.
Add to basket
Description
Description
  • Round jaw
  • Cast iron chromium plated or steel zincked
  • DIN 12894
Specifications
Specifications
1 Ea.
Zinc-plated Steel
180 mm
Clamp
80 mm
Round