Learn More
Staphylococcus Aureus Enterotoxin B Antibody (S643), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-73025AF532
 View more versions of this product
Product Code. 18569797
  • € 519.00 / 0.20mL
Define Immunology Research on Your Terms With R&D Systems
Use promo code 24090
View all promotions
Description
Staphylococcus Aureus Enterotoxin B Monoclonal antibody specifically detects Staphylococcus Aureus Enterotoxin B in Bacteria samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Staphylococcus Aureus Enterotoxin B
Monoclonal
Alexa Fluor 532
S. aureus
Hybridoma clone has been derived from hybridization of X63-Ag8-653 myeloma cells with spleen cells of Balb/c mice immunized with recombinant S. aureus enterotoxin B expressed in E. coli.
0.2 mL
Primary
Store at 4°C in the dark.
IgG1
Sandwich ELISA
S643
50mM Sodium Borate
Mouse
Protein G purified
RUO
Bacteria