missing translation for 'onlineSavingsMsg'
Learn More
Learn More
StAR Antibody (STAR/2154), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-75742CL1
This item is not returnable.
View return policy
Description
StAR Monoclonal antibody specifically detects StAR in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Protein Array
Specifications
| StAR | |
| Monoclonal | |
| CoraFluor 1 | |
| cholesterol trafficker, mitochondrial steroid acute regulatory protein, StAR, StARD1, STARD1StAR-related lipid transfer (START) domain containing 1, START domain containing 1, START domain-containing protein 1, steroid acute regulatory protein, steroidogenic acute regulator, steroidogenic acute regulatory protein, steroidogenic acute regulatory protein, mitochondrial | |
| Recombinant fragment (around aa 39-108) of human StAR protein (exact sequence is proprietary) (Uniprot: P49675) | |
| 0.1 mL | |
| Cancer | |
| 6770 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| STAR/2154 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction