missing translation for 'onlineSavingsMsg'
Learn More
Learn More
StAR Antibody (STAR/2154), Novus Biologicals™
Mouse Monoclonal Antibody
€ 277.00 - € 474.00
Specifications
| Antigen | StAR |
|---|---|
| Clone | STAR/2154 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18661799
|
Novus Biologicals
NBP2-75741-20ug |
20 μg |
€ 277.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18637386
|
Novus Biologicals
NBP2-75741-100ug |
100 μg |
€ 474.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
StAR Monoclonal antibody specifically detects StAR in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Protein ArraySpecifications
| StAR | |
| 0.2 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| Unconjugated | |
| Mouse | |
| Cancer | |
| 10 mM PBS with 0.05% BSA | |
| cholesterol trafficker, mitochondrial steroid acute regulatory protein, StAR, StARD1, STARD1StAR-related lipid transfer (START) domain containing 1, START domain containing 1, START domain-containing protein 1, steroid acute regulatory protein, steroidogenic acute regulator, steroidogenic acute regulatory protein, steroidogenic acute regulatory protein, mitochondrial | |
| Recombinant fragment (around aa 39-108) of human StAR protein (exact sequence is proprietary) (Uniprot: P49675) | |
| Primary | |
| Store at 4°C. |
| STAR/2154 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P49675 | |
| 6770 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title