Learn More
STAR Rabbit anti-Human, Mouse, Porcine, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12225-1-AP-150UL
 View more versions of this product
Product Code. 16809813
  • € 410.00 / 150µL
Description
Steroidogenic acute regulatory protein (StAR) plays a significant role in cholesterol transport from the cytoplasmic outer membrane to the inner mitochondrial membrane. The 37 kDa precursor is cleaved to generate an active 28 kDa protein capable of facilitating cholesterol metabolism into pregnenolone. StAR is prevalently expressed in mitochondria of steroid-producing adrenal and gonadal tissue. Abnormalities in StAR gene expression are impacted in autosomal Lipoid Congenial Adrenal Hyperplasia (LCAH) resulting in defects in pregnenolone and cortisol synthesis. The mechanism of cholesterol binding to StAR has yet to be elucidated.
Specifications
Specifications
STAR
Polyclonal
Unconjugated
STAR
STAR, STARD1
Rabbit
Antigen Affinity Chromatography
RUO
20845, 25557, 396597, 6770
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.34 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P49675, P51557, P97826, Q28996
STAR
STAR Fusion Protein Ag2862
150 μL
Primary
Human, Mouse, Pig, Rat