missing translation for 'onlineSavingsMsg'
Learn More

StAR (Steroidogenic Acute Regulator) (Leydig Cell Marker) Mouse, Clone: STAR/2140, enQuireBio™

Product Code. 15930928
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15930928 100 μg 100µg
15920928 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15930928 Supplier EnquireBio Supplier No. 6770MSM3P1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Specifications

Antigen StAR (Steroidogenic Acute Regulator) (Leydig Cell Marker)
Applications Immunohistochemistry
Cellular Localization Cytoplasmic (Mitochondria)
Classification Monoclonal
Clone STAR/2140
Concentration 200μg/mL
Conjugate Unconjugated
Formulation 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Host Species Mouse
Immunogen Recombinant fragment (apprx. aa 39-108) of human STAR protein
Molecular Weight of Antigen 30kDa
Positive Control K-562 cells. Adrenal or Testicular Carcinoma
Purification Method Protein A/G purified from Bioreactor concentrate.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6770
Content And Storage Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.