missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ STARD5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15662755
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15662755 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15662755 Supplier Invitrogen™ Supplier No. PA548459

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

STARD5 Polyclonal Antibody for Western Blot, IHC (P), Flow

May be involved in the intracellular transport of sterols or other lipids. May bind cholesterol or other sterols.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STARD5
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene STARD5
Gene Accession No. Q9NSY2
Gene Alias 18B7-T7(GS); 2310058G22Rik; AI607880; D7Ertd152e; RGD1561783; StAR related lipid transfer domain containing 5; Stard5; StAR-related lipid transfer (START) domain containing 5; StAR-related lipid transfer domain containing 5; stAR-related lipid transfer protein 5; StAR-related protein 1-4E; StAR-related protein p3-15E/p3-16E; START domain containing 5; START domain-containing protein 5
Gene Symbols STARD5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 26-55 amino acids from the N-terminal region of human STARD5.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80765
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.