missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ STARD7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530772
This item is not returnable.
View return policy
Description
Recommended positive controls: Jurkat. Predicted reactivity: Human (99%), Mouse (90%), Rhesus Monkey (100%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May play a protective role in mucosal tissues by preventing exaggerated allergic responses. [UniProt].
Specifications
| STARD7 | |
| Polyclonal | |
| Unconjugated | |
| STARD7 | |
| AI852671; AL022671; AW544915; gestational trophoblastic tumor protein 1; GTT1; StAR related lipid transfer domain containing 7; StARD7; StAR-related lipid transfer (START) domain containing 7; StAR-related lipid transfer domain containing 7; stAR-related lipid transfer protein 7, mitochondrial; START domain containing 7; START domain-containing protein 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56910 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9NQZ5 | |
| STARD7 | |
| Recombinant protein encompassing a sequence within the center region of human StARD7. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction