missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly6K Antibody (CL2433), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-36763-25ul
This item is not returnable.
View return policy
Description
Ly6K Monoclonal specifically detects Ly6K in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Ly6K | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| cancer/testis antigen 97, CO16, CT97, FLJ35226, HSJ001348, Ly-6K, lymphocyte antigen 6 complex, locus K, lymphocyte antigen 6K | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 54742 | |
| Protein A purified | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL2433 | |
| Western Blot 1:100-1:250, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Q17RY6 | |
| LY6K | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:TDEGDNRVWCHVCERENTFECQNPRRCKWTEPYCVIAAVKIFPRFFMVAKQCSAGCAAMERPKPEEKRFLLEEPMPFFYLKCCKIRYCNLEGPPINSSVFKEYAG | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction