missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Starter Kit for MethoCult™ H4034 Optimum
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
- The Starter Kit for MethoCult™ H4034 Optimum (MethoCult™ GF H4034) is recommended for laboratories in the initial stages of establishing procedures for assessing human hematopoietic progenitor cells using colony-forming unit (CFU) assays
- These products support the growth of clonogenic hematopoietic progenitor cells from human bone marrow, peripheral blood, cord blood, leukapheresis products and purified progenitor-enriched cells
- H4034 supports the growth of erythroid progenitors (BFU-E and CFU-E), granulocyte-macrophage progenitors (CFU-GM, CFU-G and CFU-M) and multi-potential granulocyte, erythroid, macrophage, megakaryocyte progenitors (CFU-GEMM)
- Each kit contains instructional materials in addition to all reagents and materials necessary to perform 24 duplicate assays
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells |
| For Use With (Application) | Cell Culture, Colony Assay, Functional Assay |
| Quantity | 1 Kit |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction