missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT1 Antibody (OTI4H9), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74375F
This item is not returnable.
View return policy
Description
STAT1 Monoclonal specifically detects STAT1 in Human, Mouse, Rat, Canine, Monkey, Primate samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| STAT1 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| STAT1 | |
| Full length recombinant protein of human STAT1 (NP_009330) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey, Primate | |
| IgG1 |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI4H9 | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| DKFZp686B04100, ISGF-3,91kD, signal transducer and activator of transcription 1, 91kDa, signal transducer and activator of transcription-1 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Breast Cancer, Cancer, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 6772.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction