missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT1, p Ser727 Antibody (PSM1), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-515AF488
This item is not returnable.
View return policy
Description
STAT1 Monoclonal antibody specifically detects STAT1 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| STAT1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| DKFZp686B04100, ISGF-3,91kD, signal transducer and activator of transcription 1, 91kDa, signal transducer and activator of transcription-1 | |
| STAT1 peptide sequence 721-733 (Ser727 phosphorylated). | |
| 0.1 mL | |
| Apoptosis, Breast Cancer, Cancer, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 6772 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunoprecipitation | |
| PSM1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction