missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT1, p Tyr701 Antibody (1086B), Alexa Fluor™ 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB2894G
This item is not returnable.
View return policy
Description
STAT1 Monoclonal antibody specifically detects STAT1 in Human samples. It is validated for Western Blot, Flow Cytometry
Specifications
| STAT1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry | |
| 1086B | |
| Western Blot, Intracellular Staining by Flow Cytometry | |
| DKFZp686B04100, ISGF-3,91kD, signal transducer and activator of transcription 1, 91kDa, signal transducer and activator of transcription-1 | |
| Phosphopeptide containing the human STAT1 Y701 site, Accession # NP009330 | |
| 0.1 mL | |
| Apoptosis, Breast Cancer, Cancer, Myeloid derived Suppressor Cell, Phospho Specific, Signal Transduction, Transcription Factors and Regulators | |
| 6772 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction