missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STAT1, Rabbit anti-Human, APC, Clone: 3E6, Abnova™
Rabbit Monoclonal Antibody
Brand: Abnova MAB19118.10Reactions
This item is not returnable.
View return policy
Specifications
| STAT1 | |
| Monoclonal | |
| APC | |
| In PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide). | |
| DKFZp686B04100/ISGF-3/STAT91 | |
| Rabbit | |
| 10 Reactions | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| 3E6 | |
| Flow Cytometry (5 uL/106 cells or 5 u per 100 uL) The optimal working dilution should be determined by the end user. | |
| signal transducer and activator of transcription 1, 91kDa | |
| STAT1 | |
| A synthetic phosphopeptide corresponding to residues surrounding Tyr701 of human phospho STAT1. | |
| RUO | |
| 6772 | |
| Store at 4°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction